Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Total Estrogens Double Antibody RIA Kit

Catalog No. p-7162259
Click to view available options
Quantity:
100 Tubes
500 Tubes

Quantitative Liquid Phase Radioimmunoassay

  • Total Estrogens Double Antibody RIA Kit detects the combined concentrations of unconjugated estradiol (E2) and estrone (E1).
  • Does not detect estrogen sulfates or glucuronides
  • Will not distinguish between free E1 or E2 or E2 bound to circulating binding proteins
  • Total Estrogen measurements are an approximation of plasma estrogenicity since E2 is biologically more active than E1 and the E2/E1 ratio may fluctuate widely under varying physiological and pathological conditions.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 100 Tubes
Sample Volume 500 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.