Learn More
Supply SolutionsSupplier Diversity Partner Hach Chemical - Sofchek Total Hardness; Number Of Tests: 250; Smallest Increment Steps: Steps: 0, 25, 50, 120, 250, 425 Ppm 0, 1.5, 3, 7, 15, 25 Gpg DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2793844
Total hardness test strips are easy to use, disposable, and inexpensive. It is used to obtain quick, quantitative answers in the field or in the lab. In the field, technicians can test many samples in only a few minutes, and make immediate evaluations on site. In the lab, test strips are ideal in pre test screening to detect the presence of materials that might interfere with lab testing. They are manufactured with a unique reagent pad at the tip, using proven chemistries based on standard reference methods. Test strips are available for a wide selection of parameters. Part Number 2793844: Strips are individually wrapped for maximum portability and stability.; Number of tests: 250; Smallest Increment Steps: Steps: 0, 25, 50, 120, 250, 425 ppm 0, 1.5, 3, 7, 15, 25 gpg; Footnote: mgL unless otherwise noted, ppb = ugL, ppm = mgL., gpg = grains per gallon, 1 gpg = 17.1 mgL or 17.1; Platform : Test Strip; Range: 0 to 425 ppm; Ship Wt. (lbs): 0.2; Range 2: 0 to 25 gpg; Model: Test Strips;
Catalog No. 50-476-380