Learn More
Ansh Labs Total IGFBP-4 ELISA has a total 2.5 hours incubation, calibrators and range of 6, 50-702.2 ng/mL, sensitivity of 4.735 ng/mL, sample of 25 µL Serum. Reacts with Bovine, Caprine, Equine, Human.
SDP
Supplier:  Ansh Labs AL126
Encompass_Preferred
Total IGFBP-4 ELISA has a total 2.5 hours incubation, calibrators and range of 6, 50-702.2 ng/mL, sensitivity of 4.735 ng/mL, sample of 25 µL Serum. Reacts with Bovine, Caprine, Equine, Human. Crossreactivity with related molecules is not significant.The ratio of Total (AL-126) to Intact IGFBP-4 (AL-128) concentration measured in individual subject over time will help normalizes the IGFBP-4 variability between subjects and also increase the detection rate of increased PAPP-A activity in MI subjects. The immunoassay methods designed for the measurement of total and Intact IGFBP-4 in patient samples could be of practical value for the diagnosis or prediction of various pathologies including ACS and cancer. Ansh Labs is a USA FDA registered manufacturer of diagnostic test kits, antibodies, antigens, and proteins.
Catalog No. NC2196739
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.