Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Ionic Strength Adjustment Buffer (TISAB II) with CDTA, APHA/EPA for Fluoride, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 86701/LC

Packaging Poly Natural
Quantity 4L

 View more versions of this product

Catalog No. 86701LC

  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1310-73-2,64-19-7,7647-14-5,13291-61-7
82.0,5.87,4.91,4.77,0.33
85.0,6.11,5.12,4.96,0.34
Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II), With CDTA

Specifications

7732-18-5,1310-73-2,64-19-7,7647-14-5,13291-61-7
85.0,6.11,5.12,4.96,0.34
APHA/EPA for Fluoride
Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II), With CDTA
82.0,5.87,4.91,4.77,0.33
Poly Natural
4L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit