Total Ionic Strength Adjustment Buffer (TISAB II) with CDTA, APHA/EPA for Fluoride, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 86705

Packaging Cubitainer™
Quantity 20L

 View more versions of this product

Catalog No. 8670-5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,1310-73-2,64-19-7,7647-14-5,13291-61-7
    CAS Min % 82.0,5.87,4.91,4.77,0.33
    CAS Max % 85.0,6.11,5.12,4.96,0.34
    Chemical Name or Material Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II), With CDTA

    Specifications

    Quantity 20L
    Methods APHA/EPA for Fluoride
    CAS Max % 5.12,4.96,0.34,85.0,6.11
    CAS 7647-14-5,13291-61-7,7732-18-5,1310-73-2,64-19-7
    CAS Min % 4.77,0.33,82.0,5.87,4.91
    Packaging Cubitainer™
    Chemical Name or Material Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II), With CDTA

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit