Learn More
Crystal Chem IncSupplier Diversity Partner Total Proinsulin ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 90110
The Total Proinsulin ELISA is used to quantify levels of human total intact proinsulin in serum and plasma using only a 50 µL sample volume. Highlights: Kits use a sample volume of only 50 µL. Highly specific for total proinsulinDynamic Range of 0.5 - 220 pmol/mL. Highly precise with low CV's.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7926