Learn More
Magid™ TouchMaster Lightweight Seamless Lisle Inspection Gloves
Gloves are constructed from a blend of cotton and polyester for a cool and comfortable fit. Over-edge hem ensures a secure fit, while also preventing the cuff from unraveling during laundering.
$10.08 - $12.47
Specifications
Color White
Hand Ambidextrous
Material Cotton/Polyester Blend
Product Type Inspection Gloves
Series TouchMaster 1365
View More Specs
Products 2
Catalog Number Mfr. No. Gender Price Quantity & Availability  
Catalog Number Mfr. No. Gender Price Quantity & Availability  
19-278490
View Documents
Magid™
13651
Men's
Dozen of 12 for $10.08
Case of 50 DZ for $504.50
Add to cart
19-278489
View Documents
Magid™
13650
Women's
Dozen of 12 for $12.47
Case of 50 DZ for $624.00
Add to cart
Description
Description
  • Machine-knit lisle glove fits like a dress glove
  • Seamless, reversible design for longer wear
  • Cotton/Polyester blend provides a cool and comfortable fit
  • Over-edge hem helps to prevent unraveling when laundered
  • Ideal for inspection, or any general purpose applications
  • Color-coded hems for easy size identification
Specifications
Specifications
White
Cotton/Polyester Blend
TouchMaster 1365
Ambidextrous
Inspection Gloves
Reversible