Learn More
Ansell TouchNTuff™ 83-300 Ultra-Soft Polyisoprene Gloves
Ultra-soft polyisoprene gloves offer a comfortable second skin feel for longer wear time and reduced hand fatigue.
Manufacturer:  Ansell 83300085
 View more versions of this product
Catalog No. 17-999-806
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Compatible with Class 100 (ISO 5) / Grade A Cleanroom Environments
  • Latex-free polyisoprene material offers the performance of natural rubber latex without the risk of latex sensitization
  • Ideal for double donning
  • Prevents Type I allergies
Specifications
Specifications
White
Polyisoprene
TouchNTuff™ 83-300
11.6 in.
CE 0086, Category III, EN 420:2003 + A1:2009, EN 455 1-4
Yes
Type B
Yes
Beaded
83300085
8.5
29.5 cm
ASTM D3577, ASTM D7160
Type B
Bacteria, Fungi only
ISO 10282, ISO 13485, ISO 14001, ISO 9001