Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ TouchNTuff™ 92-600 Nitrile Gloves

Ansell™ TouchNTuff™ 92-600 Nitrile Gloves offers superior splash resistance against a wide range of hazardous chemicals.

Specifications

Color Green
Material Nitrile
Powdered Powder-free
Sterility Non-sterile
For Use With (Application) Laboratory analysis, Electronic manufacture, Pharmaceuticals
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-010-412
View Documents
Ansell™
585834/DEL
7 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-010-413
View Documents
Ansell™
585835/DEL
8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-010-414
View Documents
Ansell™
585836/DEL
9 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-010-415
View Documents
Ansell™
585837/DEL
9.5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Enhanced chemical splash protection for longer periods
  • Offers superior comfort due to proprietary nitrile formulation
  • Delivers strong grip in wet or dry conditions
  • Highly versatile and suitable for many different uses
  • Silicone free for product protection
Specifications

Specifications

Green
Powder-free
Laboratory analysis, Electronic manufacture, Pharmaceuticals
Ambidextrous
Nitrile
Non-sterile
Beaded Cuff
Yes

*Warning: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit