Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
Click to view available options
Size:
6.5/7
7.5/8
8.5/9
9.5/10
Description
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
- Does not contain natural rubber latex proteins
- Strong grip in wet or dry conditions
- Rolled beaded for easy donning
- Powder free, Polymer-coated
- Ambidextrous
- Textured Fingers
- AQL: 1.5
Specifications
Chlorinated | Yes |
AQL | 1.5 |
Color | Green |
Disposable | Single-use |
Finger Thickness (English) | 6.2 mil |
Finger Thickness (Metric) | 0.16 mm |
Latex Free | Yes |
Palm Thickness (English) | 4.7 mil |
Palm Thickness (Metric) | 0.12 mm |
Size | 8.5/9 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction