Learn More
DYNAMISM INC Tough PLA Filament | .75k | 2.85mm diameter  DFS Item
Manufacturer:  DYNAMISM INC PLATOUGH
Tough PLA’s impact strength is much closer to ABS than regular PLA, so you can print parts for practical applications with heat resistance up to 60 degrees celcius.
Catalog No. NC1644337