Learn More
Brady™ ToughBond™ B-422 Polyester BMP71 Printer Labels with Permanent Adhesive
For use in commercial/industrial applications. Brady B-422 polyester label are for use with BMP71 printer.
$73.90 - $211.32
Specifications
Product Type Asset Tracking Label
Material Polyester (B-422)
Adhesive Type Acrylic, Permanent
Description Chemical, Corrosion, Humidity, UV and Weather-Resistant
Certifications/Compliance UL Recognized; CSA Accepted
View More Specs
Products 9
Catalog Number Mfr. No. Size (Metric) Size (English) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Size (English) Quantity Price Quantity & Availability  
19-108-113
View Documents
Brady™
M71C1000422
W x H: 25.4 mm x 15.2m 50 ft. x 1 in. 1 Ea.
Each for $142.46
Add to cart
19-108-118
View Documents
Brady™
M71C1900422
W x H: 38.1 mm x 15.2m 50 ft. x 1.5 in. 1 Ea.
Each for $211.32
Add to cart
19-108-121
View Documents
Brady™
M71C375422
W x H: 9.5 mm x 15.2m 50 ft. x 0.375 in. 1 Ea.
Each for $109.84
Add to cart
19-108-122
View Documents
Brady™
M71C475422
W x H: 12.06 mm x 15.2m 50 ft. x 0.475 in. 1 Ea.
Each for $119.15
Add to cart
19-108-123
View Documents
Brady™
M71C500422
W x H: 12.7 mm x 15.2m 50 ft. x 0.5 in. 1 Ea.
Each for $109.84
Add to cart
19-108-127
View Documents
Brady™
M71C625422
W x H: 15.8 mm x 15.2m 50 ft. x 0.625 in. 1 Ea.
Each for $155.51
Add to cart
19-109-099
View Documents
Brady™
M7130422
38.1 x 19 mm (W x H) 1.5 x 0.75 in. (W x H) 250/Roll
Each for $84.42
Add to cart
19-109-136
View Documents
Brady™
M7137422
76.2 x 48.2 mm (W x H) 3 x 1.9 in. (W x H) 100/Pk.
Each for $73.90
Add to cart
19-109-149
View Documents
Brady™
M7145422
38.1 x 19 mm (W x H) 1.5 x 0.75 in. (W x H) 500/Roll
Each for $90.96
Add to cart
Description
Description
Specifications
Specifications
Asset Tracking Label
Acrylic, Permanent
UL Recognized; CSA Accepted
Gloss
Component ID, Equipment ID, Barcoding, Panel ID, Voice/Data ID, Asset tracking, Rating plates, General purpose ID
Polyester (B-422)
Chemical, Corrosion, Humidity, UV and Weather-Resistant
0.004 in.
BMP™71 Printer
White