Learn More
Brady™ ToughBond™ B-483 Gloss Polyester Labels with an Aggressive Adhesive
For use in commercial/industrial applications. Brady B-483 gloss polyester labels with an aggressive adhesive is used for BMP71 printer.
$35.95 - $228.60
Specifications
Product Type Asset Tracking Label
Material Polyester (B-483)
Adhesive Type Rubber, Permanent
Description Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
Certifications/Compliance UL Recognized, CSA Approved
View More Specs
Products 13
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
19-108-119
View Documents Promotion Details
Brady™
M71C1900483
W x H: 48.3 mm x 15.2m 50 ft. x 1.9 in.
Each for $228.60
Add to cart
19-108-126
View Documents Promotion Details
Brady™
M71C500483
W x H: 12.7 mm x 15.2m 50 ft. x 0.5 in.
Each for $108.60
Add to cart
19-109-066
View Documents Promotion Details
Brady™
M7116483
25.4 x 34.9 mm (W x H) 1 x 0.375 in. (W x H)
Each for $94.41
Add to cart
19-109-073
View Documents Promotion Details
Brady™
M7119483
25.4 x 25.4 mm 1 x 1 in.
Each for $65.37
Add to cart
19-109-079
View Documents Promotion Details
Brady™
M7120483
50.8 x 25.4 mm (W x H) 2 x 1 in. (W x H)
Each for $55.21
Add to cart
19-109-084
View Documents Promotion Details
Brady™
M7123483
101.6 x 25.4 mm (W x H) 4 x 1 in. (W x H)
Each for $76.89
Add to cart
19-109-096
View Documents Promotion Details
Brady™
M7129483
38.1 x 12.7 mm (W x H) 1.5 x 0.5 in. (W x H)
Each for $106.42
Add to cart
19-109-124
View Documents Promotion Details
Brady™
M7130483
38.1 x 19.1 mm (W x H) 1.5 x 0.75 in. (W x H)
Each for $92.18
Add to cart
19-109-129
View Documents Promotion Details
Brady™
M7131483
38.1 x 25.4 mm (W x H) 1.5 x 1 in. (W x H)
Each for $79.83
Add to cart
19-109-138
View Documents Promotion Details
Brady™
M7137483
76.2 x 48.3 mm (W x H) 3 x 1.9 in. (W x H)
Each for $79.83
Add to cart
19-109-142
View Documents Promotion Details
Brady™
M7138483
101.6 x 48.3 mm (W x H) 4 x 1.9 in. (W x H)
Each for $92.18
Add to cart
19-109-151
View Documents Promotion Details
Brady™
M7161483
1.27 x 50.8 mm (W x H) 0.5 x 2 in. (W x H)
Each for $35.95
Add to cart
19-109-153
View Documents Promotion Details
Brady™
M7165483
34.9 x 25.4 mm (W x H) 1.375 x 1 in. (W x H)
Each for $57.42
Add to cart
Description
Description
Specifications
Specifications
Asset Tracking Label
Rubber, Permanent
UL Recognized, CSA Approved
Matte
Component ID, Equipment ID, Barcoding, Panel ID, Voice/Data ID, Asset tracking, Rating plates, General purpose ID
Polyester (B-483)
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
0.004 in.
BMP™71 Printer
White