Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Toughbond™ Series Patch Panel Adhesive Strip Labels, Polyester (B-422)

Configured into a continuous roll sized to a very thin strip for marking terminal blocks and patch panels, as well as general purpose cut-to-length gerneral ID applications. Brady™ Toughbond™ Series Patch Panel Adhesive Strip Labels feature a unique adhesive.

$68.50

Specifications

Finish Gloss
Material Polyester (B-422)
Color White
Thickness (English) 0.004 in.
Adhesive Type Acrylic, Permanent
View More Specs

Products 1
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19-109-027 View Documents Brady™
B33C312422
0.312 in. Each for $68.50
Add to cart
 
Description

Description

  • Feature a unique adhesive to allow labels to adhere to smooth or highly textured/powder coated surfaces
  • For use with BBP™33 Printers
  • Available in various sizes
Specifications

Specifications

Gloss
White
Acrylic, Permanent
85 ft.
Polyester (B-422)
0.004 in.
UL Recognized; CSA Accepted
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit