Learn More
Brady™ ToughWash Label Material for BMP71 Printer
For use with BMP 71 printers. Brady Toughwash material withstands harsh, aggressive environments.
$117.00 - $232.50
Specifications
Product Type ToughWash label material
Material Polyester (B-855)
Description Suggested ribbon series: R6000
Thickness (English) 0.007 in.
For Use With (Application) Safety signs and labels, Arc Flash labels, Facility identification signs and labels, Equipment labels, Visual Workplace labels
View More Specs
Products 4
Catalog Number Mfr. No. Color Width (Metric) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Width (Metric) Width (English) Price Quantity & Availability  
19-108-097 View Documents Brady™
BM71C1000855WT
White 2.54 cm 1 in. Each for $117.00
Add to cart
19-108-098 View Documents Brady™
BM71C1000855YL
Yellow 2.54 cm 1 in. Each for $117.00
Add to cart
19-108-105 View Documents Brady™
BM71C2000855WT
White 5.08 cm 2 in. Each for $232.50
Add to cart
19-108-106 View Documents Brady™
BM71C2000855YL
Yellow 5.08 cm 2 in. Each for $232.50
Add to cart
Description
Description
  • Polyester film with acrylic adhesive
  • Formulated to withstand harsh wash down process in Food and Beverage facilities
Specifications
Specifications
ToughWash label material
Suggested ribbon series: R6000
Safety signs and labels, Arc Flash labels, Facility identification signs and labels, Equipment labels, Visual Workplace labels
22.8 m
1 roll
Polyester (B-855)
0.007 in.
BMP71 printer
75 ft.