Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOX Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | TOX |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125511
|
Novus Biologicals
NBP310305100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
TOX Polyclonal specifically detects TOX in Human samples. It is validated for Western Blot.Specifications
TOX | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer | |
PBS buffer, 2% sucrose | |
9760 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
KIAA0808TOX1, thymocyte selection-associated high mobility group box, thymocyte selection-associated high mobility group box protein TOX, Thymus high mobility group box protein TOX | |
The immunogen is a synthetic peptide directed towards the middle region of Human TOX. Peptide sequence GGSNVPHNSPSPPGSKSATPSPSSSVHEDEGDDTSKINGGEKRPASDMGK | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title