Learn More
FUJIFILM Wako Toximaster™ QC8 MPR, Part 11 Software
Toximaster™ QC8 software for the Biotek ELx808IU microplate reader
Manufacturer:  FUJIFILM Wako 29035971
Catalog No. 12-333-046
Add to cart
Description
Description
  • FDA 21 CFR Part 11 ERES compliant
  • Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
  • Capable of creating three types of curves:
    • Internal standard curve
    • Manual input curve
    • Measured curve
  • Product endotoxin limit and MVD generation
  • Allows trending of product results over time and early detection of potential product failures
  • Generates hard copy printouts with all pertinent information for routine audits through three unique audit trails
  • Creation of a standard workflow to be reviewed, confirmed and approved prior to operating
  • Security functions to lock the application, disable an account and lock out the system
  • Backup of user management database and automatic backup of system information database
  • Improved precision and accuracy over traditional gel clot method
  • Ideal software for comparative testing and validating most LAL
Specifications
Specifications
Biotek ELx808IU microplate reader
Polycarbonate, Plastic
Software
FDA 21 CFR Part 11 ERES compliant, Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
Software