Learn More
FUJIFILM Wako Toximaster™ QC8 Part 11 Software PC Set
Set containing laptop computer and Toximaster™ QC8 software to be used with the Biotek ELx808IU. Instrument not included.
Manufacturer:  FUJIFILM Wako 29235931
Catalog No. 12-333-050
Description
Description
  • FDA 21 CFR Part 11 ERES compliant
  • Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
  • Capable of creating three types of curves:
    • Internal standard curve
    • Manual input curve
    • Measured curve
  • Product endotoxin limit and MVD generation
  • Allows trending of product results over time and early detection of potential product failures
  • Generates hard copy printouts with all pertinent info for routine audits through three unique audit trails
  • Creation of standard workflow to be reviewed, confirmed and approved prior to operating
  • Security functions to lock the application, disable an account and lock out the system
  • Backup of user management database and automatic backup of system info database
  • Improved precision and accuracy over traditional gel clot method
  • Ideal software for comparative testing and validating most LAL
Specifications
Specifications
Computer with Software
Biotek ELx808IU
FDA 21 CFR Part 11 ERES compliant, Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
Laptop computer and Toximaster™ QC8 software