Learn More
FUJIFILM Wako Toximaster™ QC8, Part 11 Software
Toximaster™ QC8 software for Toxinometer™ ET-7000
Manufacturer:  FUJIFILM Wako 29335961
Catalog No. 12-333-047
Add to cart
Description
Description
  • FDA 21 CFR Part 11 ERES compliant
  • Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
  • Capable of creating three types of curves:
    • Internal standard curve
    • Manual input curve
    • Measured curve
  • Product endotoxin limit and MVD generation
  • Allows trending of product results over time and early detection of potential product failures
  • Generates hard copy printouts with all pertinent information for routine audits through three unique audit trails
  • Creation of a standard workflow to be reviewed, confirmed and approved prior to operating
  • Security functions to lock the application, disable an account and lock out the system
  • Backup of user management database and automatic backup of system information database
  • Improved precision and accuracy over traditional gel clot method
  • Ideal software for comparative testing and validating most LAL
Specifications
Specifications
FDA 21 CFR Part 11 ERES compliant, Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
Software
Toxinometer™ ET-7000
Software
Polycarbonate, Plastic