Learn More
FUJIFILM Wako Toxinometer™ ET-7000/E Expansion Module
16-well expansion unit for Toxinometer™ ET-7000
Manufacturer:  FUJIFILM Wako 29235811
Catalog No. 12-333-045
Add to cart
Description
Description
  • Single assay module can simultaneously measure up to 16 samples
  • Expansion modules available (up to 8) to extend number of samples in multiples of 16
  • Single-test configuration avoids hot wells phenomenon associated with microplate readers by evenly distributing temperature in all single wells
  • Reduces chance of contamination due to single-well formats
  • Ideal for users converting from gel clot method to kinetic turbidimetric assays
  • 220 to 240 ± 10% VAC
Specifications
Specifications
Expansion Module
16-well expansion unit
For converting from gel clot method to kinetic turbidimetric assays
For Toxinometer™ ET-7000