Learn More
Medchemexpress LLC Toxoflavin | 84-82-2 | MFCD00459812 | 99.9% | 193.16 g/mol | C7H7N5O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100760100MG
Encompass_Preferred
Toxoflavin is a small-molecule research compound that modulates signaling and epigenetic targets. It acts as an antagonist of the transcription factor 4 (TCF4)/β-catenin complex and inhibits the histone demethylase KDM4A; it has reported antitumor and antibiotic activities and is supplied for research use only.
  • Antagonist of TCF4/β-catenin signaling.
  • Inhibitor of histone demethylase KDM4A.
  • Reported antitumor and antibiotic activity in research studies.
  • High chemical purity suitable for analytical and biological assays.
  • Soluble in DMSO and water; warming or sonication may aid dissolution.
Catalog No. 50-002-12473