Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Toxoplasma gondii Ab-1 Rabbt Polyclonal Antbody with BSA and azide

Toxoplasma Gondii Ab-1, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Supplier:  Epredia™ RB282R1

 View more versions of this product

Catalog No. RB282R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Toxoplasma is a crescent shaped sporozoan that lives as intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregnant women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregnancy, can produce blindness or mental retardation in the newborn.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Intact Toxoplasma gondii

Positive Control: Infected brain tissue

Cellular Localization: Whole organism

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Specifications

Specifications

Toxoplasma Gondii Ab-1
Polyclonal
Unconjugated
Intact Toxoplasma gondii
ASR
Primary
Prediluted
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
1 mL
Infectious Disease
Human
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit