Learn More
BioworldSupplier Diversity Partner Toxoplasma Gondii P30 (SAG 1) (Recombinant), 100 μg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220607281
Recombinant Toxoplasma Gondii Surface Antigen SAG-1/p30 Source: E.coli Description : The protein contains the P30 (sag1) immunodominant regions, amino acids:(45-198). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein conc: 1mg/ml in 50 mM Tris-HCl, 1.5 M urea and 50% glycerol. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of Toxoplasma gondii-infected individuals. Application: AAntigen in ELISA and Western blots, excellent antigen for detection of Toxoplasma gondii- with minimal specificity problems. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-826