Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53I13 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen TP53I13
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179466
SDP
View Documents
Novus Biologicals
NBP179466
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

TP53I13 Polyclonal specifically detects TP53I13 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TP53I13
Polyclonal
Rabbit
NP_612358
90313
Synthetic peptide directed towards the middle region of human TP53I13The immunogen for this antibody is TP53I13. Peptide sequence IYWGPTADSQDTVAAVLKRRLLQPSRRVKRSRRRPLLPPTPDSGPEGESS.
Primary
Western Blot
Unconjugated
RUO
DSCP1Damage-stimulated cytoplasmic protein 1, tumor protein p53 inducible protein 13, tumor protein p53-inducible protein 13
TP53I13
IgG
42 kDa
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit