Learn More
Aviva Systems Biology Corp TP53INP1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp AVARP09041_P050
Rabbit polyclonal antibody that recognizes the N terminal of TP53INP1(tumor protein p53 inducible nuclear protein 1) protein. It is suitable for WB application; 50ug/50ul
Catalog No. 50-014-53776