Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TP73-AS1 Monoclonal Antibody (OTI2B9)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525703

Catalog No. PIMA525703


Only null left
Add to Cart

Description

Description

TP73-AS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

TP73-AS1 in molecular biology is P73 antisense RNA 1T (non-protein coding) and also known as PDAM, is a long non-coding RNA. It may regulate apoptosis via regulation of p53-dependent anti-apoptotic genes. It may play a role in the development of oligodendroglial tumours.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TP73-AS1
Monoclonal
0.65 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
0
TP73-AS1
Full length human recombinant protein of KIAA0495.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI2B9
Unconjugated
TP73-AS1
KIAA0495; PDAM; TP73 antisense RNA 1; TP73-AS1
Mouse
Affinity Chromatography
RUO
57212
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.