Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP73-AS1 Mouse anti-Human, Clone: OTI2A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525704

Catalog No. PIMA525704


Add to cart

Description

Description

TP73-AS1 in molecular biology is P73 antisense RNA 1T (non-protein coding) and also known as PDAM, is a long non-coding RNA. It may regulate apoptosis via regulation of p53-dependent anti-apoptotic genes. It may play a role in the development of oligodendroglial tumors.
Specifications

Specifications

TP73-AS1
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UF72
TP73-AS1, KIAA0495, PDAM
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57212
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2A9
Unconjugated
TP73-AS1
Liquid
TP73-AS1
Full length human recombit protein of KIAA0495 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit