Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP73 Mouse anti-Human, Clone: 38C674.2, Abnova™

Mouse monoclonal antibody raised against human TP73.

Manufacturer:  Abnova Corporation MAB12972

Catalog No. 89-935-043


Add to cart

Description

Description

This gene encodes tumor protein p73, which is a member of the p53 family of transcription factors involved in cellular responses to stress and development. The family members include p53, p63, and p73 and have high sequence similarity to one another, which allows p63 and p73 to transactivate p53-responsive genes causing cell cycle arrest and apoptosis. The family members can interact with each other in many ways involving direct or indirect protein interactions, resulting in regulation of the same target gene promoters or regulation of each other's promoters. The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. The p73 gene expresses at least 35 mRNA variants due to the use of alternate promoters, alternate translation initiation sites, and multiple splice variations. Theoretically this can account for 29 different p73 isoforms; however, the biological validity and the full-length nature of most variants have not been determined. [provided by RefSeq]

Sequence: LYVGDPARHLAT
Specifications

Specifications

TP73
38C674.2
Mouse monoclonal antibody raised against human TP73.
In PBS (0.05% BSA, 0.05% sodium azide).
P73
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ChIP assay, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ChIP Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TP73
A synthetic peptide corresponding to amino acids 2-13 of human TP73.
Protein G purification
RUO
Primary
7161
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit