Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52L1/D53 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30327720UL

 View more versions of this product

Catalog No. NB095420


Add to cart

Description

Description

TPD52L1/D53 Polyclonal antibody specifically detects TPD52L1/D53 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

TPD52L1/D53
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunofluorescence
PBS with 50% glycerol, pH7.3.
D53, MGC8556, tumor protein D52-like 1hD53TPD52L2, tumor protein D52-like 2, tumor protein D53
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human TPD52L1/D53 (NP_001003396.1).
Affinity purified
RUO
Primary
7164
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit