Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52L1/D53 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen TPD52L1/D53
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095419
View Documents
Novus Biologicals
NBP303277100UL
100 μg Each for $320.71
Add to cart
 
NB095420
View Documents
Novus Biologicals
NBP30327720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

TPD52L1/D53 Polyclonal antibody specifically detects TPD52L1/D53 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

TPD52L1/D53
Unconjugated
Rabbit
Human
D53, MGC8556, tumor protein D52-like 1hD53TPD52L2, tumor protein D52-like 2, tumor protein D53
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human TPD52L1/D53 (NP_001003396.1).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
7164
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit