Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52L3 Mouse anti-Human, Clone: OTI4C12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526020

Catalog No. PIMA526020


Add to cart

Description

Description

Belongs to the TPD52 family and is specifically expressed in testis. Expressed at 5.6-fold higher levels in adult testis than in fetal testis.
Specifications

Specifications

TPD52L3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96J77
TPD52L3;Tumor protein D55; Tumor protein D55; Testis development protein NYD-SP25; Tumor protein D55; Tumor protein D52-like 3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
89882
Immunohistochemistry (Paraffin), Western Blot
OTI4C12
Unconjugated
TPD52L3
Liquid
TPD52L3
Full length human recombit protein of TPD52L3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit