Learn More
TPH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 208791AP
Catalog No. 50-173-3135
Add to cart
Description
Description
Tryptophan hydroxylase (TPH) catalyzes the 5-hydroxylation of tryptophan, which is the first step in the biosynthesis of indoleamines (serotonin and melatonin) (Martinez et al., 2001). In mammals, serotonin biosynthesis occurs predomitly in neurons which originate in the Raphe nuclei of the brain, and melatonin synthesis takes place within the pineal gland. Although TPH catalyzes the same reaction within the Raphe nuclei and the pineal gland, TPH activity is rate-limiting for serotonin but not melatonin biosynthesis. Serotonin functions mainly as a neurotransmitter, whereas melatonin is the principal hormone secreted by the pineal gland. The activity of TPH is enhanced by phosphorylation by cAMP-dependent protein kinase (PKA) and Ca2+/calmodulin kinase II (CAM K II) (Jiang et al., 2000; Johansen et al., 1996). Both PKA and CAM K II phosphorylate Ser58 which lies within the regulatory domain of TPH (Kuhn et al., 1997).
Specifications
Specifications
TPH1
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17532, P17752
TPH, TPH1, TPRH, TRPH, Tryptophan 5 hydroxylase 1, Tryptophan 5 monooxygenase 1, tryptophan hydroxylase 1
Rabbit
IgG
150 μL
-20°C
Primary
21990, 7166
Western Blot
Unconjugated
TPH1
Liquid
TPH1
TPH1 Fusion Protein Ag14978
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse