Learn More
TPH2 Rabbit anti-Human, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 225901AP
Catalog No. 50-173-3899
Add to cart
Description
Description
Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predomitly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
Specifications
Specifications
TPH2
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CGU9, Q8IWU9
NTPH, TPH2, Tryptophan 5 hydroxylase 2, Tryptophan 5 monooxygenase 2, tryptophan hydroxylase 2
Rabbit
IgG
150 μL
-20°C
Primary
121278, 317675
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TPH2
Liquid
Tph2
TPH2 Fusion Protein Ag18408
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat