Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPM4 Monoclonal Antibody (TM228), Invitrogen™

Catalog No. PIMA532983
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIMA532983 100 μg
1 options

Catalog No. PIMA532983

Supplier: Thermo Scientific MA532983

Mouse Monoclonal Antibody

Reconstitution information: Add 1 mL of PBS buffer will yield a concentration of 100 μg/mL.

TPM4 (tropomyosin 4) is a member of the tropomyosin family of actin-binding proteins involved in the contractile system of striated and smooth muscles and the cytoskeleton of non-muscle cells. TPM4 plays a central role in association with the troponin complex in the calcium dependent regulations of vertebrate striated muscle contraction. Tropomyosins are dimers of coiled-coil proteins that polymerize end-to-end along the major groove in most actin filaments. They provide stability to the filaments and regulate access of other actin-binding proteins. In muscle cells, they regulate muscle contraction by controlling the binding of myosin heads to the actin filament. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPM4
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone TM228
Conjugate Unconjugated
Formulation Ascites diluted in 1.2% sodium acetate with 2MG BSA, 2MG BSA and 0.01MG sodium azide, 0.01MG sodium azide
Gene TPM4
Gene Accession No. P67936
Gene Alias Tropomyosin alpha-4 chain; Tropomyosin-4; TM-4; Tpm4
Gene Symbols TPM4
Host Species Mouse
Immunogen Chicken gizzard tropomyosin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100343175, 107055375, 7171
Target Species Chicken, Human, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.