Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPM4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548443

Catalog No. PA548443

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with pig based on sequence homology.

Binds to actin filaments in muscle and non-muscle cells. Plays a central role, in association with the troponin complex, in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. In non-muscle cells is implicated in stabilizing cytoskeleton actin filaments. Binds calcium.
Specifications

Specifications

TPM4
Polyclonal
PBS with 0.09% sodium azide
P67936
Tropomyosin alpha-4 chain, TM30p1, Tropomyosin-4, TPM4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7171
Flow Cytometry, Western Blot
Unconjugated
TPM4
Liquid
TPM4
KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human TPM4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit