Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPMPA, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective, competitive GABAA-ρ antagonist with only minimal effects on GABAA and GABAB receptors (Kb values are 2.1 μM (antagonist), 320 μM (antagonist) and EC50 ∽ 500 μM (weak agonist) respectively). Displays 8-fold selectivity for human recombinant ρ1 receptors over ρ2 receptors.
Specifications
Quantity | 10 mg |
Formula Weight | 161.14 |
Percent Purity | >98% |
Chemical Name or Material | TPMPA |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction