Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPMT Mouse anti-Human, DyLight 488, Clone: 2E7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197939G
Description
TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
TPMT | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TPMT | |
Recombinant human TPMT (1-245aa) purified from E. coli | |
100 μL | |
Proteases & Other Enzymes, Signal Transduction | |
7172 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
20000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human TPMT mAb, clone 2E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TPMT protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title