Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPPP/p25 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19161320UL

 View more versions of this product

Catalog No. NBP19161320


Add to cart

Description

Description

TPPP/p25 Polyclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

TPPP/p25
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
TPPP
Synthetic peptide directed towards the C terminal of mouse 2900041A09RIK. Peptide sequence KAVSSPTVSRLTDTSKFTGSHKERFDQSGKGKGKAGRVDLVDESGYVPGY.
Protein A purified
RUO
Primary
11076
Western Blot
Western Blot 1:1000
NP_878259
p24, P24,25 kDa brain-specific protein, P25, p25alpha, p25-alpha, TPPP/p25brain specific protein p25 alpha, TPPP1glycogen synthase kinase 3 (GSK3) inhibitor p24, tubulin polymerization promoting protein, tubulin polymerization-promoting protein
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit