Learn More
TPPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 150571AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9939
Add to cart
Description
Description
This antibody detects the 19-20 kDa TPPP3 in HeLa and PC-3 cells. The non-specific bands between 40 and 60 kDa were also occasionally observed.
TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.
Specifications
Specifications
TPPP3
Polyclonal
Unconjugated
TPPP3
CGI 38, p20, p25gamma, TPPP/p20, TPPP3
Rabbit
Antigen Affinity Chromatography
RUO
291966, 51673, 67971
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5PPN5, Q9BW30, Q9CRB6
TPPP3
TPPP3 Fusion Protein Ag7040
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG