Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPRA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534275
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Opossum (100%).
TPRA1 gene ontology (GO) annotations include aging; embryonic cleavage; lipid metabolic process; negative regulation of mitotic cell cycle phase transition.
Specifications
TPRA1 | |
Polyclonal | |
Unconjugated | |
TPRA1 | |
40kDa; FLJ32197; G protein-coupled receptor 175; GPR175; Integral membrane protein GPR175; PP6566; seven transmembrane domain orphan receptor; TMEM227; TPRA1; Tpra40; Transmembrane domain protein of 40 kDa regulated in adipocytes; transmembrane domain protein regulated in adipocytes 40 kDa; transmembrane protein 227; transmembrane protein adipocyte associated 1; transmembrane protein adipocyte-associated 1; transmembrane protein, adipocyte asscociated 1; transmembrane protein, adipocyte associated 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131601 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q86W33 | |
TPRA1 | |
Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human TPRA1 / GPR175. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction