Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPRG1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120409

This item is not returnable.
View return policy
Description
Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Cytoplasmic vesicle, secretory vesicle, synaptic vesicle.
TPRG1L Polyclonal antibody specifically detects TPRG1L in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TPRG1L | |
Polyclonal | |
Unconjugated | |
Tprgl | |
1200015A19Rik; Fam79a; family with sequence similarity 79, member A; h-mover; mossy fiber terminal-associated vertebrate-specific presynaptic protein; mossy-fiber terminal-associated vertebrate-specific presynaptic protein; Mover; Protein FAM79A; RP11-46F15.3; Svap30; synaptic vesicle associated protein of 30 kD; Tprg1l; Tprgl; transformation related protein 63 regulated like; tumor protein p63 regulated 1-like; tumor protein p63-regulated gene 1-like protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human TPRG1L (NP_877429.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
3.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A8WCF8, Q9DBS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67808, 687090 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title