Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPSG1 Mouse anti-Human, Clone: OTI1G1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525837

Catalog No. PIMA525837


Add to cart

Description

Description

Tryptase gamma, also known as serine protease 31 or transmembrane tryptase, is an enzyme that in humans is encoded by the TPSG1 gene. Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. There is uncertainty regarding the number of genes in this cluster. Currently four functional genes -alpha I, beta I, beta II and gamma I - have been identified. And beta I has an allelic variant named alpha II, beta II has an allelic variant beta III, also gamma I has an allelic variant gamma II. Beta tryptases appear to be the main isoenzymes expressed in mast cells, whereas in basophils, alpha-tryptases predomit. This gene differs from other members of the tryptase gene family in that it has C-terminal hydrophobic domain, which may serve as a membrane anchor. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders.
Specifications

Specifications

TPSG1
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRR2
Tryptase gamma, Tryptase gamma, Tryptase- gamma, Serine protease 31, Transmembrane tryptase, Tryptase gamma light chain, Tryptase gamma heavy chain, PRSS31, TMT
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25823
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1G1
Unconjugated
TPSG1
Liquid
TPSG1
Human recombit protein fragment corresponding to amino acids 20-283 of TPSG1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit