Learn More
TPTE2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251201AP
Catalog No. 50-173-4734
Add to cart
Description
Description
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002]
Specifications
Specifications
TPTE2
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6XPS3
Lipid phosphatase TPIP, TPIP, TPTE2
Rabbit
IgG
150 μL
-20°C
Primary
93492
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TPTE2
Liquid
TPTE2
TPTE2 Fusion Protein Ag19169
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human