Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPX2 Mouse anti-Human, Clone: TPX2-01, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267265
Description
TPX2 Monoclonal antibody specifically detects TPX2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
TPX2 | |
TPX2-01 | |
Unconjugated | |
Purified | |
C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis) | |
TPX2 | |
Human TPX2 | |
Monoclonal | |
>95% by SDS-PAGE | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators | |
Human,Mouse,Rat |
ELISA,Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot | |
1mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry | |
PBS (pH 7.4) with 15mM Sodium Azide | |
22974 | |
Mouse | |
IgG1 | |
Primary | |
0.1mg | |
Store at 4°C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title