Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPX2 Mouse anti-Human, DyLight 755, Clone: TPX2-01, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267265IR

Catalog No. NBP267265IR



Description

Description

TPX2 Monoclonal antibody specifically detects TPX2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

TPX2
TPX2-01
DyLight 755
Purified
C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis)
TPX2
Human TPX2
Monoclonal
>95% by SDS-PAGE
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators
Human,Mouse,Rat
Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry
1mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry
50MM Sodium Borate with 0.05% Sodium Azide
22974
Mouse
IgG1
Primary
0.1mL
Store at 4°C in the dark.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit