Learn More
MilliporeSigma™ TRA-1-60, Clone: TRA-1-60
Supplier: MilliporeSigma™ MAB4360/DEL

Description
This antibody reacts with TRA-1-60 antigen that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is seen with murine EC, EG or ES cells.Both the TRA-1-60 and TRA-1-81 monoclonal antibodies (MAB4381) recognize antigens that are associated with a pericellular matrix proteoglycan. TRA-1-60 reacts with a sialidase-sensitive epitope whilst TRA-1-81 reacts with an unknown epitope of the same molecule.
Andrews, P W, et al(1984). Three monoclonal antibodies defining distinct differentiation antigens associated with different high molecular weight polypeptides on the surface of human embryonal carcinoma cells.Hybridoma 3: 347-61
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications
TRA-1-60 | |
1.18mg/mL | |
Mouse | |
100 μg | |
Stem Cell Biology | |
Human | |
IgM |
Monoclonal | |
Unconjugated | |
Human embryonal carcinoma cell line 2102Ep | |
RUO | |
Primary | |
Semi-purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.