Learn More
TRA2B Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 238321AP
Catalog No. 50-173-4133
Add to cart
Description
Description
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
Specifications
Specifications
TRA2B
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62995, P62996
Htra2 beta, SFRS10, SRFS10, TRA 2 beta, TRA2 BETA, TRA2B, TRAN2B
Rabbit
IgG
150 μL
-20°C
Primary
20462, 6434
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRA2B
Liquid
TRA2B
TRA2B Fusion Protein Ag20683
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse