Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAAK Mouse anti-Mouse,Rat, Unconjugated, Clone: N445/27 , MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen TRAAK
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN23931MI
View Documents
MilliporeSigma
MABN2393100UG
100 μg
Each for $372.00
Add to cart
 
MABN23932MI
View Documents
MilliporeSigma
MABN239325UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

TRAAK Monoclonal antibody specifically detects TRAAK in Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

TRAAK
Unconjugated
Mouse
Potassium channel subfamily K member 4;TWIK-related arachidonic acid-stimulated potassium channel protein;TRAAK
A recombinant fragment corresponding to 143 amino acids from the C-terminal, cytoplasmic domain of mouse Potassium channel subfamily K member 4 (TRAAK).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin)
Purified
Mouse, Rat
Kcnk4, Traak
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit