Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEXCertiPrep™ Trace Metals on Cellulose Ester Filter Paper

Matrix-matched standards designed for the needs of various types of industrial hygiene and air monitoring programs.

$105.00 - $120.00

Specifications

Autoclavable No
Bubble Point ≥ 0.1 bar, air with water at 23°C
Diameter (Metric) Outer 37 mm
Flow Rate 190 mL/min x cm2 (Water) | 16 L/min x cm2 (Air)
For Use With (Application) Aerosol monitoring
View More Specs

Products 7
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
CFMSPIKE1
View Documents
SPEXCertiPrep™
CFMSPIKE1
15 Low-level trace metals
Each for $105.00
Add to cart
 
CFMSPIKE1U
View Documents
SPEXCertiPrep™
CFMSPIKE1U
16 Low-level trace metals; With Uranium
Each for $120.00
Add to cart
 
CFMSPIKE2
View Documents
SPEXCertiPrep™
CFMSPIKE2
15 Mid-level trace metals
Each for $105.00
Add to cart
 
CFMSPIKE2U
View Documents
SPEXCertiPrep™
CFMSPIKE2U
16 Mid-level trace metals; With Uranium
Each for $120.00
Add to cart
 
CFMSPIKE3
View Documents
SPEXCertiPrep™
CFMSPIKE3
15 High-level trace metals
Each for $105.00
Add to cart
 
CFMSPIKE4
View Documents
SPEXCertiPrep™
CFMSPIKE4
16 Low-level trace metals; With Aluminum
Each for $116.00
Add to cart
 
CFMSPIKE5
View Documents
SPEXCertiPrep™
CFMSPIKE5
16 Mid-level trace metals; With Aluminum
Each for $120.00
Add to cart
 
Description

Description

  • Lab control samples for method development, validating laboratory performance, proficiency test validation, and quality control checks
  • Ideal for measuring sample preparation and analysis accuracy and precision
  • Formulated to meet testing requirements for NIOSH 7300-Series, Standards for ASTM D7035, ISO 15202-1 and 15202-2, and CEN (EN) 13890. 
  • ISO/IEC 17025 and ISO 17034 Accredited
  • ISO 9001 Certified
Specifications

Specifications

No
37 mm
Aerosol monitoring
0.8 µm
≥ 0.1 bar, air with water at 23°C
190 mL/min x cm2 (Water) | 16 L/min x cm2 (Air)
Mixed Cellulose Esters (MCE), Cellulose Pad
10 Spiked Filters and 5 Blanks
SDS

For Laboratory Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit