Learn More
Thermo Scientific™ TRACE™ TR-1701 GC Columns Available on GSA/VA Contract for Federal Government customers only.
Use mid-polarity columns with alternative selectivity with Thermo Scientific™ TRACE™ TR-1701 GC Columns.
Manufacturer:  Thermo Scientific™ 260Q131P
 View more versions of this product
Catalog No. 26-0Q1-31P
  Check Availability
Add to cart
Specifications
Description
Use mid-polarity columns with alternative selectivity with Thermo Scientific™ TRACE TR-1701 GC Columns.
  • Mid-polarity phase, 14% Cyanopropylphenyl polysiloxane
  • Low bleed even at a high operating temperature
  • Excellent starting point for method development
  • Suitable for a wide variety of applications
  • Similar to DB-1701, Rtx-1701, HP-1701, BP10, OV-1701, 007-1701, CP-Sil 19 CB
Recommended for:
  • Pesticides
  • PCB's
  • PAH's
  • Organic Acids
  • Drugs
  • Steroids
  • EPA 608, 8081
Description & Specifications
Specifications
0.32mm
280/300°C
G46
15m
15m x 0.32mm x 0.25um
GC/MS
0.32mm
0.25μm
Trace TR-1701